missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Vinyl-1-cyclohexene, 97%, stabilized, Thermo Scientific Chemicals
10.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 mL
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -101.0°C |
Color | Beige-Brown to White |
Density | 0.8320g/mL |
Boiling Point | 126.0°C to 127.0°C |
Flash Point | 16°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- 4-Vinyl-1-cyclohexene, stabilized
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H412-Harmful to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P331-Do NOT induce vomiting.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'