missing translation for 'onlineSavingsMsg'
Learn More

4-Vinylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 10412985
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2156-04-9
Molecular Formula C8H9BO2
Molecular Weight (g/mol) 147.97
MDL Number MFCD00239441
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 190°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 16, IV, 1676
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,dmso,dmf,thf,diethyl ether
IUPAC Name (4-ethenylphenyl)boronic acid
PubChem CID 2734393
Formula Weight 147.97
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Vinylbenzeneboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Vinylbenzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'