missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Vinylbenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in dichloromethane and methanol
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
It is used as a metabolite of 1,4-diethenylbenzene. It is also an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.
Solubility
Soluble in dichloromethane and methanol
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼142°C |
Density | 1.3 |
Quantity | 25 g |
Beilstein | 2041130 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in dichloromethane and methanol |
Formula Weight | 148.16 |
Percent Purity | 98% |
Chemical Name or Material | 4-Vinylbenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'