missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Vinylbenzyl chloride, 90%, technical, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 88.0 |
CAS Max % | 100.0 |
Color | Yellow |
Density | 1.0800g/mL |
Flash Point | 105°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 88% min. (GC) |
Packaging | Glass bottle |
Linear Formula | CH2CHC6H4CH2Cl |
Refractive Index | 1.5731 to 1.5751 |
Show More |
Safety and Handling


Product Identifier
- 4-Vinylbenzyl chloride
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P331-Do NOT induce vomiting.
- P363-Wash contaminated clothing before reuse.
- P403-Store in a well-ventilated place.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 4-Vinylbenzyl chloride
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Vinylbenzyl chloride, 90%, technical, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction