Learn More
Thermo Scientific™ 4-Way Flipper™ Racks
GREENER_CHOICE
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™4-Way Flipper™ Racks.
Brand:  Thermo Scientific™ 8860
421.69 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11520125
  • £607.00 / Pack of 20
Estimated Shipment: 12-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hold thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side, using Thermo Scientific™ 4-Way Flipper™ Racks.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4-way Flipper Rack
Fluorescent Orange
Thirty-two 0.5mL, thirty-two 1.5mL, twelve 15mL or four 50mL tubes, each on a different side
4-Way Flipper Rack
0.5 mL, 15 mL, Tubes (50 mL)
Rectangular
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ 4-Way Flipper™ Racks > 4-Way Flipper Rack, Fluorescent Orange