Learn More
Masterflex™ 4-way Stopcock
Click to view available options
Connectors:
Male Luer Lock
Male Luer Slip
Unit Size:
Pack of 10
Description
Use these stopcocks for a variety of applications. Cole-Parmer™ 4-way Stopcocks feature a PC body and an HDPE stem.
- Four-way stopcock
- Choose stopcocks with either a male luer lock or a male luer slip
- Handle rotates 360 degrees on four-way stopcocks
Specifications
Specifications
| Type | Stopcock |
| Material | Polycarbonate, HDPE |
| Plug Material | Polycarbonate and HDPE |
| Outlet Port | Male Luer Lock or A Male Luer Slip |
| Connectors | Male Luer Lock |