Learn More
TubeWriter 400 - Custom Rack Side Print for Any Labware
Custom racks made to exact specifications to print directly on side of unique labware
Brand:  TubeWriter PS-0000XX-EL-400-Custom
Product Code. 30067431
  • £2640.00 / Each
Explore more special offers
Description
Description
  • Custom racks desinged for the 400
  • Racks made to your exact specifications to directly print and label on the side of any labware
  • Rack will ensure correct alignment and holds labware securely
  • No time-consuming sticky labels or handwriting required
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Specifications
Labeling Rack