Learn More
TubeWriter 400 - Custom Rack Top Print for Any Labware
Custom racks made to exact specifications to print directly on the top of unique labware
Brand:  TubeWriter PT-000XX-EL-400-Custom
Product Code. 30067428
  • £2640.00 / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Custom racks designed for the 400
  • Racks made to your exact specifications to directly on the top of any labware
  • Rack will ensure correct alignment and holds labware securely
  • No time-consuming sticky labels or handwriting required
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Specifications
Specifications
Labeling Rack
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TubeWriter 400 - Custom Rack Top Print for Any Labware > Custom Rack Top Print for Any Labware