Learn More
RETSCH 400 dia. x 65mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
£273.00 - £440.00
Specifications
Diameter (Metric) Outer | 400 mm |
---|---|
Frame Material | Stainless Steel |
Mesh Material | Stainless Steel |
For Use With (Application) | For separation, fractioning, particle size determination |
Quantity | 1Each |
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Price | Quantity & Availability | |||||
10024981
|
RETSCH
60.167.001400 |
1.40mm |
£273.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10488341
|
RETSCH
60.167.001180 |
1.18mm |
£273.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10428731
|
RETSCH
60.167.001700 |
1.7mm |
£273.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10075061
|
RETSCH
60.167.100000 |
100mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10503732
|
RETSCH
60.167.106000 |
106mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10640462
|
RETSCH
60.167.125000 |
125mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10249071
|
RETSCH
60.167.012500 |
12.50mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10196311
|
RETSCH
60.167.011200 |
11.20mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10670102
|
RETSCH
60.167.013200 |
13.2mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10136461
|
RETSCH
60.167.019000 |
19mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10488151
|
RETSCH
60.167.001000 |
1mm |
£273.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10660452
|
RETSCH
60.167.016000 |
16mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10583912
|
RETSCH
60.167.002800 |
2.8mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10024991
|
RETSCH
60.167.002360 |
2.36mm |
£273.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10497961
|
RETSCH
60.167.022400 |
22.40mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10196501
|
RETSCH
60.167.025000 |
25mm |
£386.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10438541
|
RETSCH
60.167.026500 |
26.50mm |
£425.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10438731
|
RETSCH
60.167.002000 |
2mm |
£273.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10749512
|
RETSCH
60.167.031500 |
31.50mm |
£425.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10228501
|
RETSCH
60.167.037500 |
37.50mm |
£425.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10468921
|
RETSCH
60.167.003350 |
3.35mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10186071
|
RETSCH
60.167.004750 |
4.75mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10054981
|
RETSCH
60.167.045000 |
45mm |
£425.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10476041
|
RETSCH
60.167.004000 |
4mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10390952
|
RETSCH
60.167.005600 |
5.6mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10720283
|
RETSCH
60.167.050000 |
50mm |
£425.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10730283
|
RETSCH
60.167.053000 |
53mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10446621
|
RETSCH
60.167.006300 |
6.3mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10770853
|
RETSCH
60.167.006700 |
6.7mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10553342
|
RETSCH
60.167.063000 |
63mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10640102
|
RETSCH
60.167.008000 |
8mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10740283
|
RETSCH
60.167.075000 |
75mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10034911
|
RETSCH
60.167.009500 |
9.5mm |
£325.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
10620292
|
RETSCH
60.167.090000 |
90mm |
£440.00
Each |
Estimated Shipment: 14-10-2024
Log in to see stock. |
|||||
Specifications
400 mm | |
Stainless Steel | |
1Each | |
ASTM E 11 | |
Shaker AS 450 |
Stainless Steel | |
For separation, fractioning, particle size determination | |
Test Sieve | |
Stainless Steel | |
65mm |