Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ISO Certified
Brand:  RETSCH 60.166.000038
 View more versions of this product
Product Code. 10258581
  • £580.00 / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
400 mm
Stainless Steel
Test Sieve
Stainless Steel
65mm
For separation, fractioning, particle size determination
Stainless Steel
38 μm
ISO 3310/1
Shaker AS 450