missing translation for 'onlineSavingsMsg'
Learn More
Learn More
400 NTU Ratio Turbidity Standard, Reagecon

Suspension of extremely accurate (± 1%) polymer microspheres with guaranteed shelf life stability even after opening the bottle.
Brand: Reagecon CRSR-400-100
This item is not returnable.
View return policy
Description
- Turbidity Reference Standard, Turbidity Reference Material.
- Designed and formulated to overcome the adverse safety, accuracy and stability issues experienced with other turbidity standards, primarily formazin-based standards.
- Certified
- NIST traceable
- Non-toxic, non-carcinogenic
- Highly accurate
- Stable
- EPA approved
Specifications
Ratio Instruments | |
Accuracy ± 1% NTU | |
100 mL |
Liquid format in HDPE Bottle | |
400 NTU ± 1% |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
3 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
CRSR40024B1 | Certificate of Analysis | 26/02/2025 | CRSR-400-100 |
CRSR40022H1 | Certificate of Analysis | 15/02/2023 | CRSR-400-100 |
CRSR40021B1 | Certificate of Analysis | 21/09/2021 | CRSR-400-100 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
400 NTU Ratio Turbidity Standard, Reagecon > 100 mL
Spot an opportunity for improvement?Share a Content Correction