missing translation for 'onlineSavingsMsg'
Learn More

LMS™ Series 3 Model 400WA Cooled Incubator, 450 L, Stainless Steel

Temperature Range: -10°C to +50°C

Brand:  LMS™ 400A

Product Code. 12863456

  • £7985.00 / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Range of mid-sized benchtop and freestanding cooled incubators with a white stove enamel mild steel exterior, foam insulation and stainless steel interiors
  • Reliable and accurate temperature control
  • Standard single and dual temperature, plus programmable models available
  • Temperature range -10.0°C to +50.0°C, temperature variation with average workload±0.5°C
  • Self-evaporating condensate drip tray
  • White plastic coated wire shelves
  • Energy efficient control system and eco-friendly
  • P.I.D. heating and cooling to ensure stable internal temperature
  • Pre-set safety cutout
  • Hermetically sealed refrigeration system
  • Door lock
  • Digital controls are easy to read and simple to use
  • Full range offactory-fitted optional extras available, including LED lighting, access porthole, alarm system, inner perspex door, internal socket and chart recorder
  • Ideal for applications such as growth studies, B.O.D.tests, seed germination and vernalisation, tissue culture, enzymology, serum studies and immunology, sample storage or product shelf life tests
Specifications

Specifications

ISO 9001:2008
Freestanding incubator
Stainless Steel
510 mm x 600 mm
Stove Enameled Mild Steel
-10°C to +50°C
Digital
400WA
CFC-free
±0.5°C
27.1 in.
690 mm
71.2 in.
1810 mm
27.6 in.
140 kg
230 V
Dual temperature
450 L
Fan assisted air circulation via premixing chamber
Optional Fluorescent/UV
3
Refrigerated Incubator
Foamed polyurethane
1
±0.1°C
2 Years
21.6 in.
550 mm
46.1 in.
1170 mm
700 mm
240 V, 50 Hz
50 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.