missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Clamp-On Breaker Lockout, 480/600V

For use with single-pole and internal-trip multi-pole breakers

Brand:  Brady™ 065966

 View more versions of this product

Product Code. 15673507

  • £72.80 / Pack of 6
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made of rugged polypropylene and impact modified nylon
  • Accepts lock shackles up to 0.28 in. in diameter
  • Includes Cceats for use with breakers with long, sliding switch throws

Specifications

Specifications

Clamp-on Breaker Lockout
Nylon
0.28 in.
2.175 x 1.588 x 3.253 in.
Cleats
6/Pk.
Red
UL 94 HB
0.7 cm
40.30 mm
Clamp-on
40.30 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.