missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Refrigerator/Freezer Ultra™ Thermometer

Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.

Brand:  Fisherbrand™ 93677-93

 View more versions of this product

Product Code. 15527593

  • £104.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips

Description

Description

Monitor two different areas at the same time with Traceable™ Refrigerator/Freezer Ultra™ Thermometer.

  • Designed to meet current CDC requirements for vaccine storage and monitoring
  • Triple display simultaneously shows min., max. and current temperatures for a specified probe
  • Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
  • Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
  • °C/°F switchable
  • Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
  • Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
  • Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
  • Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Range: -50° to +70°C (-58° to +158°F)
  • Accuracy: ±0.5°C between -30° to +50°C, otherwise ±1°C
  • Resolution: 0.1°

Certifications

Individually serial-numbered Traceable™ Certificate is provided from ISO/IEC 17025:2005 calibration laboratory accredited by A2LA; indicates traceability to standards provided by NIST.

Specifications

Specifications

Nontoxic Glycol
NIST Traceable
Monitoring temperatures in freezers, water baths, heating blocks, incubators, and refrigerators
7 cm
One Bullet Probe
4 oz.
Refrigerator/Freezer Ultra Thermometer
158°F
-58°F
-58°F to +158°F
Yes
±0.5°C (-30°C to +50°C), ±1°C otherwise
2.5 x 6.3 cm (1 x 2.5 in.)
7 x 10.8 x 2 cm (2.75 x 4.25 x 0.75 in.)
Traceable™ Certificate, flip-open stand, 10 ft. micro cable, wall mounting, Velcro™, magnetic strips
ABS Plastic
0.1°
113.4 g
3 in.
70°C
-50°C
-50°C to +70°C
Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator
Digital
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.