missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ Microplate Lids

Product Code. p-4954209 Shop All Brand Products
Click to view available options
Plate Compatibility:
1536-well Standard Plate
384-well Plate
96-well Plate
96-well Standard Plate
No. per Pack:
50
100
Condensation Rings:
No
Yes
Height (Metric):
4.5 mm
5.5 mm
8 mm
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy

Product Code. 10337483

Brand: BRAND™ 782150

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

CERTIFIED LIFE SCIENCE QUALITY plate covers for protection of samples, reaction batches, or cell cultures from contamination and evaporation.

  • Made from crystal-clear polystyrene
  • Cut-corner sets orientation of lid
  • Stackable

Lids with condensation rings available for 96-well standard plates

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Material Polystyrene
Sterility Non-sterile
No. of Wells 96
Height (Metric) 8 mm
Condensation Rings Yes
No. per Pack 100
Evaporation Barrier No
Product Type Microplate Cover
Adhesive None
Temperature Range (Metric) <-20°C
Well Shape All well shapes
Plate Compatibility 96-well Standard Plate
Cut-off Corners Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.