missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ 1000 Series Gastight™ Saltline Syringes
For use with solutions that have a high salt concentration
£144.00 - £185.00
Specifications
Autoclavable | Not Autoclavable |
---|---|
Product Type | Gastight Saltline Syringe |
Connection Type | Bubble Free Prime (BFP) |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15192514
|
Hamilton™
208336 |
5 mL |
£185.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15102524
|
Hamilton™
208337 |
10 mL |
£168.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15182514
|
Hamilton™
208335 |
1 mL |
£144.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Equipped with inert and non-corrosive plunger
- Fitted with resistant ultra-high molecular weight poly-ethylene (UHMWPE) plunger tip
Applications:
For life science applications with salty buffers, mining applications with acids or trace metal analysis
Specifications
Not Autoclavable | |
Gastight Saltline Syringe | |
Bubble Free Prime (BFP) |