Learn More
BRAND™ BRANDplates™ Insert System Microplates - 24-Well Standard Plate
24-well standard plate
Brand:  BRAND™ 782890
97.45 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.61800kg
 View more versions of this product
Product Code. 15503783
  • £128.00 / Pack of 10
In Stock
to see stock.
Add to basket
Description
Description
  • Transparent
  • Available with pureGrade™ S and cellGrade™ plus surface
  • Includes 24 individually fillable wells that can be fitted with strips of 4 inserts and/or individual inserts
  • Guide ridges for perfect insert fit
  • Designed feeding ports for easy media changes, suitable for robotics/automation
  • 10 plates/Pk. (Individually wrapped with lid)
Applications:
Co-culture, secretion studies, migration studies
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Specifications
Sterile
Cell Culture Insert System
Co-culture, secretion studies, migration studies
cellGrade plus
24
Insert system