Learn More
BRAND™ BRANDplates™ Insert System Microplates - 24-Well Standard Plate
24-well standard plate
Brand: BRAND™ 782880
131.10 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.61400kg
Description
- Transparent
- Available with pureGrade™ S and cellGrade™ plus surface
- Includes 24 individually fillable wells that can be fitted with strips of 4 inserts and/or individual inserts
- Guide ridges for perfect insert fit
- Designed feeding ports for easy media changes, suitable for robotics/automation
- 10 plates/Pk. (Individually wrapped with lid)
Applications:
Co-culture, secretion studies, migration studies
Compliance
ANSI/SLAS standards 1 and 4
Specifications
Sterile | |
Cell Culture Insert System | |
Co-culture, secretion studies, migration studies |
pureGrade S | |
24 | |
Insert system |