missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Gastight Microliter™ Syringes: RN Termination, No Needle Included

Brand:  Hamilton™ 7661-01

 View more versions of this product

Product Code. 10737453

  • £131.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • PTFE coating from point for 0.75 in.
  • For dispensing both liquids and gases
  • Precision-machined polymer plunger tip that creates leak-free seal

Available as replacement needle for installing into standard removable needle (RN) syringe; Needles sold separately

Specifications

Specifications

Autoclavable
26s G
1805
50 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers