missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Biohazard Identification Signs: Biohazard Infectious Agent Entry Requirements Contact Phone
For identification of equipment, rooms and materials that contain, or are contaminated with, hazardous agents
£27.72 - £41.98
Specifications
Height (English) | 14 in. |
---|---|
Height (Metric) | 4.2 m |
Width (English) | 10 in. |
Width (Metric) | 3 m |
Displayed Text | BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___ |
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
16421481
|
Brady™
126647 |
Polyester |
£32.85
Each |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
16411481
|
Brady™
126646 |
Plastic |
£27.72
Each |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
16401481
|
Brady™
126645 |
Aluminum |
£41.98
Each |
Estimated Shipment: 03-10-2024
Log in to see stock. |
|||||
Description
- Made from aluminum, plastic or self-sticking polyester
- Size: 14 in. H x 10 in. W
- Legend: BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___
Specifications
14 in. | |
10 in. | |
BIOHAZARD INFECTIOUS AGENT___ ENTRY REQUIREMENTS ___ CONTACT ___ PHONE ___ |
4.2 m | |
3 m | |
Biohazard Identification Sign |