missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 6 1000 D MWCO Standard RC Pre-wetted Dialysis Kits
With Pre-wetted ready-to-use tubing, not requiring soaking to remove glycerol
£141.00 - £204.00
Specifications
MWCO | 1 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1 meter of Spectra/Por 6 Dialysis Tubing
- 1 Standard Spectra/Por Closure (for buoyancy)
- 1 Weighted Spectra/Por Closure (for orientation)
- 5 Tubing Opening Picks
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15330742
|
Spectrum™
132638T |
24 mm | 4.6 mL/cm vol./length |
£198.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15320742
|
Spectrum™
132636T |
11.5 mm | 1.1 mL/cm vol./length |
£141.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15340742
|
Spectrum™
132640T |
29 mm | 6.4 mL/cm vol./length |
£204.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Clear, flexible, study standard grade Regenerated Cellulose (RC) dialysis membrane
- Membrane packaged wet in preservative solution of 0.05% sodium azide
- Kits available in 1m tubing lengths ideal for membrane evaluation and qualification
- Include a pair of closures and convenient tubing opening picks
Specifications
1 kDa | |
1 m Tubing/Kit |