missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 1000 D MWCO Standard RC Pre-Treated Dialysis Kits
With chemically-treated tubing to remove any trace levels of heavy metals and sulfide compounds
£159.00 - £280.00
Specifications
MWCO | 1 kDa |
---|---|
Quantity | 1 m Tubing/Kit |
Includes:
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
15300692
|
Spectrum™
132105T |
29 mm | 6.4 mL/cm vol./length |
£280.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15390682
|
Spectrum™
132104T |
24 mm | 4.6 mL/cm vol./length |
£159.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15380682
|
Spectrum™
132103T |
11.5 mm | 1.1 mL/cm vol./length |
£266.00
1m |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
1 kDa | |
1 m Tubing/Kit |