missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ 4B Chemical Protection Suit with Feet
8.07 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- PP spunbonded non-woven fabric with PE film
- Breathable, lightweight and flexible material for exceptional wearer comfort
- Orange taped seams for optimal protection and good visibility
- Middle finger loops prevent the sleeves from riding up
- Optimum fit for a high level of wearer comfort
- Secure and comfortable fastenings thanks to elasticated bands on hood, arms and legs
- Tailored boot sock for extra safety
- Particle-tight and spray-tight
- Anti-static properties
- Protection against infectious diseases
- Protection against pesticides
- Free of silicone, plasticizers and substances that interfere with wetting agents
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Material | Polypropylene |
Color | White, Orange |
Disposable | Single-use |
Sterility | Non-sterile |
Certifications/Compliance | Chemical protection/PPE category III, EN 14605:2005+A1:2009 (type 4), EN ISO 13982-1:2004+A1:2010 (type 5) , EN 13034:2005+A1:2010 (type 6), EN 1149-5:2008, EN 14126:2003+AC:2004, EN 1073-2:2002, DIN 32781:2010 |
For Use With (Application) | Controlled Environments |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'