missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 167.19 |
Percent Purity | 97% |
Chemical Name or Material | 4H-Thieno[3, 2-b]pyrrole-5-carboxylic acid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4H-Thieno[3,2-b]pyrrole-5-carboxylic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction