missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4M KCl Gel Saturated Refilling Solutions, Cole-Parmer™
Click to view available options
Quantity:
125 mL
Packaging:
2Each
Specifications
Specifications
Concentration | 4 M |
Quantity | 125 mL |
Formulation | 4M KCL |
Packaging | 2Each |
Safety and Handling
Product Identifier
- fill solution
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- H400-Very toxic to aquatic life.
- MIXTURE LIST-Contains : potassium chloride
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'