missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(4S)-(-)-4-Isopropyl-2-oxazolidinone, 98%, Thermo Scientific Chemicals
£67.40 - £769.00
Chemical Identifiers
CAS | 17016-83-0 |
---|---|
Molecular Formula | C6H11NO2 |
Molecular Weight (g/mol) | 129.16 |
MDL Number | MFCD00010847 |
InChI Key | YBUPWRYTXGAWJX-RXMQYKEDSA-N |
Synonym | s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon |
PubChem CID | 7157133 |
IUPAC Name | (4S)-4-propan-2-yl-1,3-oxazolidin-2-one |
SMILES | CC(C)C1COC(=O)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10397331
|
Thermo Scientific Acros
297860010 |
1 g | Glass Bottle |
£67.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10574921
|
Thermo Scientific Acros
297860050 |
5 g | Glass Bottle |
£206.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686904
|
Thermo Scientific Acros
297860250 |
25 g | Glass Bottle |
£769.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17016-83-0 | |
129.16 | |
YBUPWRYTXGAWJX-RXMQYKEDSA-N | |
7157133 | |
CC(C)C1COC(=O)N1 |
C6H11NO2 | |
MFCD00010847 | |
s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon | |
(4S)-4-propan-2-yl-1,3-oxazolidin-2-one |
Specifications
70°C to 73°C | |
97.5 | |
White | |
C6H11NO2 | |
MFCD00010847 | |
Solubility in water: insoluble in water | |
CC(C)C1COC(=O)N1 | |
-18 | |
7157133 | |
98% | |
Crystalline Powder |
17016-83-0 | |
100.0 | |
97.5% min. (GC) | |
Authentic | |
s-4-isopropyl-2-oxazolidinone, s---4-isopropyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinone, s-4-isopropyloxazolidin-2-one, 4s-4-isopropyl-1,3-oxazolidin-2-one, 2-oxazolidinone, 4-1-methylethyl-, 4s, 4s-4-propan-2-yl-1,3-oxazolidin-2-one, 4s---isopropyl-2-oxazolidinone, 4 s-1-methylethyl-2-oxazolidinone, 4s---4-isopropyl-2-oxazolidinon | |
YBUPWRYTXGAWJX-RXMQYKEDSA-N | |
(4S)-4-propan-2-yl-1,3-oxazolidin-2-one | |
129.16 | |
129.16 | |
−18° (20°C c=1,ethanol) | |
(4S)-(-)-4-Isopropyl-2-oxazolidinone |
RUO – Research Use Only