missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-[(1,3-Dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 255.23 |
Percent Purity | 97% |
Chemical Name or Material | 5-[(1, 3-Dioxo-1, 3-dihydro-2H-isoindol-2-yl)methyl]-2-furaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'