missing translation for 'onlineSavingsMsg'
Learn More

5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine, 95%

Product Code. 10751531 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 10751531

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25440-14-6 | C44H10F20N4 | 974.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25440-14-6
CAS Min % 94.0
CAS Max % 100.0
Color Purple or Violet
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C44H10F20N4
MDL Number MFCD00010032
Quantity 100 mg
Synonym 5,10,15,20-tetrakis pentafluorophenyl porphyrin, 5,10,15,20-tetrakis pentafluorophenyl-21h,23h-porphine, meso-tetra pentafluorophenyl porphine, 21h,23h-porphine, 5,10,15,20-tetrakis pentafluorophenyl, 5,10,15,20-tetrakis 2,3,4,5,6-pentafluorophenyl-21,22-dihydroporphyrin, 2,7,12,17-tetrakis 2,3,4,5,6-pentafluorophenyl-21,22,23,24-tetraazapentacyclo 16.2.1.1 3 ,?.1?, 1 1 .1 1 3 , 1 ? tetracosa-1,3,5,7,9,11 23 ,12,14,16,18 21 ,19-undecaene, acmc-209gjw, iron iii ionophore iv, tetra pentafluorophenyl poephine, tetrakis pentafluorophenyl porphyrin
InChI Key GBAPBSUXEFFUAD-UHFFFAOYSA-N
SMILES FC1=C(F)C(F)=C(C(F)=C1F)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
IUPAC Name 5,10,15,20-tetrakis(2,3,4,5,6-pentafluorophenyl)-21,22-dihydroporphyrin
Molecular Weight (g/mol) 974.56
PubChem CID 3671775
Formula Weight 974.57
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine
Show More Show Less
compliance-icons
Product Identifier
  • 5,10,15,20-Tetrakis(pentafluorophenyl)-21H,23H-porphine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.