missing translation for 'onlineSavingsMsg'
Learn More

5-(2,6-Dichlorophenyl)-1H-tetrazole, 97%, Thermo Scientific™

Product Code. 11314136
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11314136

Brand: Thermo Scientific Alfa Aesar L19258.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50907-31-8
Molecular Formula C7H4Cl2N4
Molecular Weight (g/mol) 215.04
MDL Number MFCD00808036
InChI Key RLXQNTWJPTUTCL-UHFFFAOYSA-N
Synonym 5-2,6-dichlorophenyl-1h-tetrazole, 5-2,6-dichlorophenyl-2h-tetrazole, 5-2,6-dichlorophenyl-1h-1,2,3,4-tetrazole, 5-2,6-dichlorophenyl-1h-1,2,3,4-tetraazole, maybridge1_003804, acmc-20ama2, 5-2,6-dichlorophenyl-1h-tetraazole, 1h-tetrazole, 5-2,6-dichlorophenyl

Specifications

Melting Point 203°C to 209°C
Quantity 250 mg
Beilstein 5268246
Formula Weight 215.04
Percent Purity 97%
Chemical Name or Material 5-(2,6-Dichlorophenyl)-1H-tetrazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.