missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Bromophenyl)-1,3-oxazole, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 224.06 |
Percent Purity | 95% |
Chemical Name or Material | 5-(2-Bromophenyl)-1, 3-oxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'