missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Bromophenyl)-3-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Synonym | 5-2-bromophenyl-3-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-2-bromophenyl-3-methyl, 5-2-bromophenyl-3-methyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-2-bromophenyl-3-methyl |
Formula Weight | 239.07 |
Percent Purity | 97% |
Chemical Name or Material | 5-(2-Bromophenyl)-3-methyl-1, 2, 4-oxadiazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'