missing translation for 'onlineSavingsMsg'
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Thermo Scientific™

Product Code. 10049151
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-70-0
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
InChI Key CQCKNGVVCKWDLF-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methanol, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-4-thiazolyl thiophene-2-methanol, 5-2-methylthiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl thien-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methan-1-ol
PubChem CID 2776470
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
SMILES CC1=NC(=CS1)C2=CC=C(S2)CO

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 211.31
Percent Purity 95%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'