missing translation for 'onlineSavingsMsg'
Learn More

5-[2-Methyl-3-(trifluoromethyl)-1H-2lambda∼5∼-pyrazol-5-yl]-2-thiophenecarboxylic acid, 97%, Thermo Scientific™

Product Code. p-3881274
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10022314

Brand: Thermo Scientific Maybridge MO07338DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 223499-20-5
Molecular Formula C10H7F3N2O2S
Molecular Weight (g/mol) 276.233
MDL Number MFCD00206674
InChI Key MGYVPTDPOYVOQK-UHFFFAOYSA-N
Synonym 5-1-methyl-5-trifluoromethyl pyrazol-3-yl thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl thiophene-2-carboxylic acid, 3-5-carboxythien-2-yl-1-methyl-5-trifluoromethyl-1h-pyrazole, 2-thiophenecarboxylicacid, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl, 5-1-methyl-5-trifluoromethyl-1h-pyrazol-3-yl-thiophene-2-carboxylic acid, 5-1-methyl-5-trifluoromethyl-1h-pyrazole-3-yl-2-thiophenecarboxylic acid
PubChem CID 2775669
IUPAC Name 5-[1-methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-2-carboxylic acid
SMILES CN1C(=CC(=N1)C2=CC=C(S2)C(=O)O)C(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 276.24
Percent Purity 97%
Chemical Name or Material 5-[1-Methyl-5-(trifluoromethyl)-1H-pyrazol-3-yl]thiophene-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.