missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(2-Phenylethynyl)nicotinonitrile, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS | 845266-26-4 |
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Molecular Formula | C14H8N2 |
MDL Number | MFCD01763953 |
Quantity | 1 g |
Synonym | 5-2-phenylethynyl nicotinonitrile, 5-2-phenylethynyl pyridine-3-carbonitrile, maybridge1_004883, 5-phenylethynylnicotinonitrile, 5-phenylethynyl nicotinonitrile, 5-2-phenylethynyl-3-pyridinecarbonitrile, 3-pyridinecarbonitrile,5-2-phenylethynyl |
InChI Key | SLUQJZCMAPASFJ-UHFFFAOYSA-N |
SMILES | C1=CC=C(C=C1)C#CC2=CC(=CN=C2)C#N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'