Learn More
5-(3-Chlorophenyl)-2-furoyl chloride, Thermo Scientific™
£49.40 - £177.00
Chemical Identifiers
CAS | 92973-26-7 |
---|---|
Molecular Formula | C11H6Cl2O2 |
Molecular Weight (g/mol) | 241.07 |
MDL Number | MFCD02258034 |
InChI Key | DBONYOXUBWINEQ-UHFFFAOYSA-N |
Synonym | 5-3-chlorophenyl furan-2-carbonyl chloride, 5-3-chlorophenyl-2-furoyl chloride, 5-3-chloro-phenyl-furan-2-carbonyl chloride, acmc-20am9e, 2-furancarbonylchloride, 5-3-chlorophenyl |
PubChem CID | 3854521 |
IUPAC Name | 5-(3-chlorophenyl)furan-2-carbonyl chloride |
SMILES | ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
92973-26-7 | |
241.07 | |
DBONYOXUBWINEQ-UHFFFAOYSA-N | |
3854521 | |
ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1 |
C11H6Cl2O2 | |
MFCD02258034 | |
5-3-chlorophenyl furan-2-carbonyl chloride, 5-3-chlorophenyl-2-furoyl chloride, 5-3-chloro-phenyl-furan-2-carbonyl chloride, acmc-20am9e, 2-furancarbonylchloride, 5-3-chlorophenyl | |
5-(3-chlorophenyl)furan-2-carbonyl chloride |
Spezifikation
92973-26-7 | |
MFCD02258034 | |
5-3-chlorophenyl furan-2-carbonyl chloride, 5-3-chlorophenyl-2-furoyl chloride, 5-3-chloro-phenyl-furan-2-carbonyl chloride, acmc-20am9e, 2-furancarbonylchloride, 5-3-chlorophenyl | |
ClC(=O)C1=CC=C(O1)C1=CC(Cl)=CC=C1 | |
241.07 | |
241.08 |
C11H6Cl2O2 | |
UN3261 | |
DBONYOXUBWINEQ-UHFFFAOYSA-N | |
5-(3-chlorophenyl)furan-2-carbonyl chloride | |
3854521 | |
5-(3-Chlorophenyl)-2-furoyl chloride |
Sicherheit und Handhabung
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures