missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Methoxyphenyl)-2-furoic acid, 97%, Thermo Scientific™

Product Code. 10460584
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 54022-96-7
Molecular Formula C12H10O4
Molecular Weight (g/mol) 218.21
MDL Number MFCD05022489
InChI Key RIIFZAPFLSKUTL-UHFFFAOYSA-N
Synonym 5-3-methoxyphenyl-2-furoic acid, 5-3-methoxyphenyl furan-2-carboxylic acid, 2-furancarboxylic acid, 5-3-methoxyphenyl, 2-furancarboxylic acid,5-3-methoxyphenyl, 5-3-methoxy-phenyl-furan-2-carboxylic acid
PubChem CID 2794680
IUPAC Name 5-(3-methoxyphenyl)furan-2-carboxylic acid
SMILES COC1=CC=CC(=C1)C1=CC=C(O1)C(O)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 218.21
Percent Purity 97%
Chemical Name or Material 5-(3-Methoxyphenyl)-2-furoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(3-Methoxyphenyl)-2-furoic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.