missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Pyridinyloxy)-2-furoic acid, ≥97%, Thermo Scientific™

Product Code. 10148694
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10148694

Brand: Thermo Scientific Maybridge AW01246DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-39-3
Molecular Formula C10H7NO4
Molecular Weight (g/mol) 205.169
MDL Number MFCD03407385
InChI Key ZTYMSWYWSCYFAF-UHFFFAOYSA-N
Synonym 5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acid
PubChem CID 7060545
IUPAC Name 5-pyridin-3-yloxyfuran-2-carboxylic acid
SMILES C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 205.17
Percent Purity 97%
Chemical Name or Material 5-(3-Pyridinyloxy)-2-furoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.