missing translation for 'onlineSavingsMsg'
Learn More

5-(3-Trifluoromethoxyphenyl)-1H-tetrazole, 97%, Thermo Scientific™

Product Code. 11308236
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216144-09-1
Molecular Formula C8H5F3N4O
Molecular Weight (g/mol) 230.15
MDL Number MFCD01075683
InChI Key YKYWVXHVSVSQOX-UHFFFAOYSA-N
Synonym 5-3-trifluoromethoxy phenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-2h-tetrazole, 5-3-trifluoromethoxyphenyl-1h-tetrazole, 5-3-trifluoromethoxy phenyl-1h-1,2,3,4-tetrazole, 3-1h-1,2,3,4-tetraazol-5-yl phenoxy trifluoromethane, acmc-1cs77, 5-3-trifluoromethoxy phenyl tetrazole, 2h-tetrazole,5-3-trifluoromethoxy phenyl, 5-3-trifluoromethyloxy phenyl-2h-1,2,3,4-tetrazole
PubChem CID 2777350
IUPAC Name 5-[3-(trifluoromethoxy)phenyl]-2H-tetrazole
SMILES C1=CC(=CC(=C1)OC(F)(F)F)C2=NNN=N2

Specifications

Melting Point 156°C to 159°C
Quantity 500 mg
Formula Weight 230.15
Percent Purity 97%
Chemical Name or Material 5-(3-Trifluoromethoxyphenyl)-1H-tetrazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'