Learn More
5-(4-Chlorophenyl)-2-furoic acid, 95+%, Thermo Scientific Chemicals
£33.30 - £343.29
Chemical Identifiers
CAS | 41019-45-8 |
---|---|
Molecular Formula | C11H6ClO3 |
Molecular Weight (g/mol) | 221.62 |
MDL Number | MFCD00216587 |
InChI Key | XIPQHWUSDHTXOO-UHFFFAOYSA-M |
PubChem CID | 726693 |
SMILES | [O-]C(=O)C1=CC=C(O1)C1=CC=C(Cl)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10629194
|
Thermo Scientific Acros
375540010 |
1 g |
£33.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10096452
|
Thermo Scientific Acros
375540050 |
5 g |
£110.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11926171
|
Thermo Scientific Acros
375540250 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
41019-45-8 | |
221.62 | |
XIPQHWUSDHTXOO-UHFFFAOYSA-M | |
[O-]C(=O)C1=CC=C(O1)C1=CC=C(Cl)C=C1 |
C11H6ClO3 | |
MFCD00216587 | |
726693 |
Specifications
41019-45-8 | |
Brown | |
95+% | |
C11H6ClO3 | |
5-4-chlorophenyl-2-furoic acid, 5-4-chlorophenyl furan-2-carboxylic acid, 2-furancarboxylic acid, 5-4-chlorophenyl, 5-4-chloro-phenyl-furan-2-carboxylic acid, 2-furancarboxylic acid,5-4-chlorophenyl, acid, 62, acmc-1an70, 5-p-chlorophenyl furoic acid, 5-4-chlorophenyl furoic acid, 5-p-chlorophenyl-2furoic acid | |
XIPQHWUSDHTXOO-UHFFFAOYSA-M | |
5-(4-chlorophenyl)furan-2-carboxylic acid | |
726693 | |
95+% | |
5-(4-Chlorophenyl)-2-furoic acid |
198.0°C to 201.0°C | |
Authentic | |
Glass bottle | |
MFCD00216587 | |
Solubility in water: soluble. Other solubilities: ethanol,acetone,dmf,dioxane | |
[O-]C(=O)C1=CC=C(O1)C1=CC=C(Cl)C=C1 | |
221.62 | |
222.63 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only