missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Chlorophenyl)furfural, 95+%, Thermo Scientific™

Product Code. 10230443
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 34035-03-5
Molecular Formula C11H7ClO2
Molecular Weight (g/mol) 206.63
MDL Number MFCD00195947
InChI Key ROJGJNINTRCMBL-UHFFFAOYSA-N
Synonym 5-4-chlorophenyl furan-2-carbaldehyde, 5-4-chlorophenyl-2-furaldehyde, 5-4-chlorophenyl furfural, 5-4-chloro-phenyl-furan-2-carbaldehyde, 2-formyl-5-4-chlorophenyl furan, 2-furancarboxaldehyde, 5-4-chlorophenyl, 5-4-chlorophenyl-2-furancarboxaldehyde, 5-4-chlorophenyl furan-2-carboxaldehyde, pubchem4001, cambridge id 7179578
PubChem CID 676184
IUPAC Name 5-(4-chlorophenyl)furan-2-carbaldehyde
SMILES ClC1=CC=C(C=C1)C1=CC=C(O1)C=O

Specifications

Melting Point 128°C to 131°C
CAS Min % 95.0
Color Brown
CAS Max % 100.0
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Needles
Assay Percent Range 95% min. (GC)
Formula Weight 206.63
Percent Purity 95+%
Chemical Name or Material 5-(4-Chlorophenyl)furfural
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.