Learn More
5-(4-Methoxyphenyl)-1,3-oxazole-4-carboxylic acid, 97%, Thermo Scientific™
£35.70 - £268.00
Chemical Identifiers
CAS | 89205-07-2 |
---|---|
Molecular Formula | C11H9NO4 |
Molecular Weight (g/mol) | 219.196 |
MDL Number | MFCD03644147 |
InChI Key | XVXCIDWYZMBWJU-UHFFFAOYSA-N |
Synonym | 5-4-methoxyphenyl-1,3-oxazole-4-carboxylic acid, 5-4-methoxyphenyl oxazole-4-carboxylic acid, 5-4-methoxyphenyl-1,3-oxazole-4-carboxylicacid, 4-oxazolecarboxylicacid, 5-4-methoxyphenyl, 5-4-methoxy-phenyl-oxazole-4-carboxylic acid, maybridge3_004367, acmc-20c54g, 5-4-methoxyphenyl-4-oxazolecarboxylic acid, 5-4-methoxyphenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-4-methoxyphenyl |
PubChem CID | 2779782 |
IUPAC Name | 5-(4-methoxyphenyl)-1,3-oxazole-4-carboxylic acid |
SMILES | COC1=CC=C(C=C1)C2=C(N=CO2)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10181262
|
Thermo Scientific Maybridge
MO00452CB |
250 mg |
£35.70
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10427242
|
Thermo Scientific Maybridge
MO00452DA |
1 g |
£71.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10782017
|
Thermo Scientific Maybridge
MO00452DE |
5 g |
£268.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
89205-07-2 | |
219.196 | |
XVXCIDWYZMBWJU-UHFFFAOYSA-N | |
2779782 | |
COC1=CC=C(C=C1)C2=C(N=CO2)C(=O)O |
C11H9NO4 | |
MFCD03644147 | |
5-4-methoxyphenyl-1,3-oxazole-4-carboxylic acid, 5-4-methoxyphenyl oxazole-4-carboxylic acid, 5-4-methoxyphenyl-1,3-oxazole-4-carboxylicacid, 4-oxazolecarboxylicacid, 5-4-methoxyphenyl, 5-4-methoxy-phenyl-oxazole-4-carboxylic acid, maybridge3_004367, acmc-20c54g, 5-4-methoxyphenyl-4-oxazolecarboxylic acid, 5-4-methoxyphenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-4-methoxyphenyl | |
5-(4-methoxyphenyl)-1,3-oxazole-4-carboxylic acid |
Specifications
89205-07-2 | |
97% | |
C11H9NO4 | |
5-4-methoxyphenyl-1,3-oxazole-4-carboxylic acid, 5-4-methoxyphenyl oxazole-4-carboxylic acid, 5-4-methoxyphenyl-1,3-oxazole-4-carboxylicacid, 4-oxazolecarboxylicacid, 5-4-methoxyphenyl, 5-4-methoxy-phenyl-oxazole-4-carboxylic acid, maybridge3_004367, acmc-20c54g, 5-4-methoxyphenyl-4-oxazolecarboxylic acid, 5-4-methoxyphenyl-oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-4-methoxyphenyl | |
COC1=CC=C(C=C1)C2=C(N=CO2)C(=O)O | |
219.196 | |
219.2 | |
5-(4-Methoxyphenyl)-1, 3-oxazole-4-carboxylic acid |
97.0 | |
Amber glass bottle | |
MFCD03644147 | |
XVXCIDWYZMBWJU-UHFFFAOYSA-N | |
5-(4-methoxyphenyl)-1,3-oxazole-4-carboxylic acid | |
2779782 | |
97% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.