missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Bromo-2-thienyl)isoxazole, ≥97%, Thermo Scientific™

Product Code. 10664765
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 138716-31-1
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.079
MDL Number MFCD03618406
InChI Key FCUZGSQWFAXBSB-UHFFFAOYSA-N
Synonym 2-bromo-5-isoxazol-5-yl thiophene, 5-5-bromo-2-thienyl isoxazole, 5-5-bromothiophen-2-yl-1,2-oxazole, 5-bromo-2-isoxazol-5-ylthiophene, 5-5-bromothien-2-yl isoxazole, 5-5-bromo-2-thiophenyl isoxazole, 5-5-bromothiophen-2-yl isoxazole, 5-5-bromothien-2-yl-1,2-oxazole, 5-5-bromanylthiophen-2-yl-1,2-oxazole
PubChem CID 2757026
IUPAC Name 5-(5-bromothiophen-2-yl)-1,2-oxazole
SMILES C1=C(SC(=C1)Br)C2=CC=NO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 230.08
Percent Purity 97%
Chemical Name or Material 5-(5-Bromo-2-thienyl)isoxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.