missing translation for 'onlineSavingsMsg'
Learn More

5-(5-Bromo-2-thienyl)isoxazole, ≥97%, Thermo Scientific™

Product Code. 10672055
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 138716-31-1
Molecular Formula C7H4BrNOS
Molecular Weight (g/mol) 230.079
MDL Number MFCD03618406
InChI Key FCUZGSQWFAXBSB-UHFFFAOYSA-N
Synonym 2-bromo-5-isoxazol-5-yl thiophene, 5-5-bromo-2-thienyl isoxazole, 5-5-bromothiophen-2-yl-1,2-oxazole, 5-bromo-2-isoxazol-5-ylthiophene, 5-5-bromothien-2-yl isoxazole, 5-5-bromo-2-thiophenyl isoxazole, 5-5-bromothiophen-2-yl isoxazole, 5-5-bromothien-2-yl-1,2-oxazole, 5-5-bromanylthiophen-2-yl-1,2-oxazole
PubChem CID 2757026
IUPAC Name 5-(5-bromothiophen-2-yl)-1,2-oxazole
SMILES C1=C(SC(=C1)Br)C2=CC=NO2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 230.08
Percent Purity 97%
Chemical Name or Material 5-(5-Bromo-2-thienyl)isoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'