missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Dimethylhydantoin, 97%, Thermo Scientific Chemicals
£27.10 - £247.00
Chemical Identifiers
CAS | 77-71-4 |
---|---|
Molecular Formula | C5H8N2O2 |
Molecular Weight (g/mol) | 128.13 |
MDL Number | MFCD00005266 |
InChI Key | YIROYDNZEPTFOL-UHFFFAOYSA-N |
Synonym | 5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin |
PubChem CID | 6491 |
IUPAC Name | 5,5-dimethylimidazolidine-2,4-dione |
SMILES | CC1(C)NC(=O)NC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10500581
|
Thermo Scientific Acros
116312500 |
250 g | Plastic bottle |
£27.10
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10418070
|
Thermo Scientific Acros
116310010 |
1 kg | Plastic bottle |
£57.50
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10474535
|
Thermo Scientific Acros
116310051 |
5 kg | Plastic drum |
£247.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
77-71-4 | |
128.13 | |
YIROYDNZEPTFOL-UHFFFAOYSA-N | |
6491 | |
CC1(C)NC(=O)NC1=O |
C5H8N2O2 | |
MFCD00005266 | |
5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin | |
5,5-dimethylimidazolidine-2,4-dione |
Specifications
77-71-4 | |
100.0 | |
White | |
Authentic | |
C5H8N2O2 | |
24, 289 | |
5,5-dimethylhydantoin, dimethylhydantoin, dm hydantoin, dantoin dmh, 2,4-imidazolidinedione, 5,5-dimethyl, 5,5-dimethyl-2,4-imidazolidinedione, dantoin 736, hydantoin, 5,5-dimethyl, 5,5-dimethylhydantoin dmh, 5,5-dimethyl hydantoin | |
YIROYDNZEPTFOL-UHFFFAOYSA-N | |
5,5-dimethylimidazolidine-2,4-dione | |
6491 | |
97% | |
5, 5-Dimethylhydantoin |
96.0 | |
175.0°C to 180.0°C | |
0.25% max. (60°C, 2 hrs, 1mm Hg) | |
97% | |
MFCD00005266 | |
07,126 | |
Solubility in water: soluble in water. Other solubilities: soluble in alcohol and ether,soluble in ethanol,ethylether,aceton | |
CC1(C)NC(=O)NC1=O | |
128.13 | |
128.13 | |
Crystalline Powder or Crystals |
Safety and Handling
missing translation for 'einecsNumber' : 201-051-3
RUO – Research Use Only