missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,5-Diphenylhydantoin sodium salt, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 295°C to 298°C |
Color | White |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Physical Form | Crystalline Powder |
Quantity | 25 g |
Assay Percent Range | 99% |
Merck Index | 15,7433 |
Solubility Information | Solubility in water: practically insoluble |
IUPAC Name | sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide |
Show More |
Safety and Handling
Product Identifier
- 5,5-Diphenylhydantoin sodium salt
- Warning
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'