missing translation for 'onlineSavingsMsg'
Learn More
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine dihydrochloride, 95%, Thermo Scientific™ Product Code.: 15264328

5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine dihydrochloride, 95%, Thermo Scientific™

Product Code. 15264328
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15264328

Brand: Thermo Scientific Acros 454200010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 157327-49-6
Infrared Spectrum Authentic
Assay Percent Range 94.0 to max. 106% min. (Argentometry)
Packaging Glass Bottle
Molecular Formula C7H9N3·2ClH
Quantity 1 g
Synonym 5,6,7,8-tetrahydropyrido 4,3-d pyrimidine dihydrochloride, 5h,6h,7h,8h-pyrido 4,3-d pyrimidine dihydrochloride, 5,6,7,8-tetrahydro-pyrido 4,3-d pyrimidine dihydrochloride, 5,6,7,8-tetrahydropyrido 4,3-d pyrimidine 2hcl, 5,6,7,8-tetrahydropyrido 4,3-d pyrimidinedihydrochloride, 5,6,7,8-tetrahydropyrido 4,3-d pyrimidine di, hcl
InChI Key WLSBRWTXCPUJMN-UHFFFAOYSA-N
SMILES C1CNCC2=CN=CN=C21.Cl.Cl
IUPAC Name 5,6,7,8-tetrahydropyrido[4,3-d]pyrimidine;dihydrochloride
Molecular Weight (g/mol) 208.09
PubChem CID 19883247
Formula Weight 208.09
Percent Purity 95%
Chemical Name or Material 5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 5,6,7,8-Tetrahydropyrido¢4,3-d!pyrimidine dihydrochloride, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine dihydrochloride, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.