Learn More
Thermo Scientific Chemicals 5(6)-Carboxyfluorescein
£25.20 - £123.00
Chemical Identifiers
CAS | 72088-94-9 |
---|---|
Molecular Formula | C42H24O14 |
Molecular Weight (g/mol) | 752.64 |
MDL Number | MFCD00151081 |
InChI Key | BPVHBBXCESDRKW-UHFFFAOYSA-N |
Synonym | 5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid |
PubChem CID | 44119975 |
IUPAC Name | 3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid |
SMILES | OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10269590
|
Thermo Scientific Chemicals
404105000 |
500 mg | Glass bottle |
£25.20
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10516081
|
Thermo Scientific Chemicals
404100050 |
5 g | Glass bottle |
£123.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72088-94-9 | |
752.64 | |
BPVHBBXCESDRKW-UHFFFAOYSA-N | |
44119975 | |
OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2 |
C42H24O14 | |
MFCD00151081 | |
5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid | |
3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid |
Specifications
72088-94-9 | |
99% | |
MFCD00151081 | |
Solubility in water: slightly soluble. Other solubilities: soluble in 0.1 n naoh | |
OC(=O)C1=CC2=C(C=C1)C1(OC2=O)C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2.OC(=O)C1=CC2=C(C=C1)C(=O)OC21C2=C(OC3=C1C=CC(O)=C3)C=C(O)C=C2 | |
752.64 | |
376.32 |
275°C | |
C42H24O14 | |
5 6-fam, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid compound with 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-6-carboxylic acid 1:1, 5-and-6-fam, 5-and-6-carboxyfluorescein, 5-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, dye content 90 %, 5 6-carboxyfluorescein; carboxyfluorescein, 5 6-carboxyfluorescein, bioreagent, suitable for fluorescence hplc, 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-5-carboxylic acid compound with 3,6-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9-xanthene-6-carboxylic acid 1:1, 4 5-carboxyfluorescein; 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5 6-carboxylic acid | |
BPVHBBXCESDRKW-UHFFFAOYSA-N | |
3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-carboxylic acid;3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid | |
44119975 | |
5(6)-Carboxyfluorescein, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 276-331-1
missing translation for 'tsca' : TSCA
- 5(6)-Carboxyfluorescein
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only