missing translation for 'onlineSavingsMsg'
Learn More

5,6-Diaminouracil sulfate, 95%, Thermo Scientific Chemicals

Product Code. 11936541
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42965-55-9
Molecular Formula C4H8N4O6S
Molecular Weight (g/mol) 240.19
MDL Number MFCD00135845
InChI Key IKARJSDZQCSEJX-UHFFFAOYSA-N
Synonym 5,6-diaminopyrimidine-2,4-diol sulfate, 5,6-diaminopyrimidine-2,4-diol sulphate, diaminopyrimidine-2,4-diol; sulfuric acid, 4,5-diamino-2,6-dihydroxypyrimidine sulfate, 5,6-diaminopyrimidine-2,4 1h,3h-dione sulfate, 5,6-diaminouracil; sulfuric acid, 5?6-diaminouracil sulfate, c4h6n4o2.h2so4, 5,4-dihydroxypyrimidine sulfate, bis 5,6-diaminouracil sulphate
PubChem CID 319643
SMILES OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White to Brown
Elemental Analysis 18.80 to 21.20% C, 21.93 to 24.73% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 50 g
IUPAC Name 5,6-diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione; sulfuric acid
Formula Weight 240.2
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5, 6-Diaminouracil sulfate
Show More Show Less
compliance-icons
Product Identifier
  • 5,6-Diaminouracil sulfate, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'