missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5,6-Dimethylbenzimidazole, 99+%, Thermo Scientific Chemicals
£33.60 - £108.00
Chemical Identifiers
CAS | 582-60-5 |
---|---|
Molecular Formula | C9H10N2 |
Molecular Weight (g/mol) | 146.19 |
MDL Number | MFCD00005603 |
InChI Key | LJUQGASMPRMWIW-UHFFFAOYSA-N |
Synonym | 5,6-dimethylbenzimidazole, dimedazol, dimedazole, dimesol, dimezol, 5,6-dimethyl-1h-benzo d imidazole, dimethylbenzimidazole, 1h-benzimidazole, 5,6-dimethyl, benzimidazole, 5,6-dimethyl, 5.6-dimethylbenzimidazole |
PubChem CID | 675 |
ChEBI | CHEBI:15890 |
IUPAC Name | 5,6-dimethyl-1H-1,3-benzodiazole |
SMILES | CC1=C(C)C=C2N=CNC2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10540741
|
Thermo Scientific Acros
115980250 |
25 g | Glass bottle |
£33.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306750
|
Thermo Scientific Acros
115981000 |
100 g | Glass bottle |
£108.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
582-60-5 | |
146.19 | |
LJUQGASMPRMWIW-UHFFFAOYSA-N | |
675 | |
5,6-dimethyl-1H-1,3-benzodiazole |
C9H10N2 | |
MFCD00005603 | |
5,6-dimethylbenzimidazole, dimedazol, dimedazole, dimesol, dimezol, 5,6-dimethyl-1h-benzo d imidazole, dimethylbenzimidazole, 1h-benzimidazole, 5,6-dimethyl, benzimidazole, 5,6-dimethyl, 5.6-dimethylbenzimidazole | |
CHEBI:15890 | |
CC1=C(C)C=C2N=CNC2=C1 |
Specifications
582-60-5 | |
100.0 | |
Beige | |
99+% | |
MFCD00005603 | |
5,6-dimethylbenzimidazole, dimedazol, dimedazole, dimesol, dimezol, 5,6-dimethyl-1h-benzo d imidazole, dimethylbenzimidazole, 1h-benzimidazole, 5,6-dimethyl, benzimidazole, 5,6-dimethyl, 5.6-dimethylbenzimidazole | |
LJUQGASMPRMWIW-UHFFFAOYSA-N | |
5,6-dimethyl-1H-1,3-benzodiazole | |
675 | |
146.19 | |
Powder |
99.0 | |
203°C to 206°C | |
Authentic | |
C9H10N2 | |
15, 3258 | |
Solubility in water: soluble. Other solubilities: freely soluble in diluted acids,soluble in chloroform,ether and ethanol | |
CC1=C(C)C=C2N=CNC2=C1 | |
146.19 | |
CHEBI:15890 | |
99+% | |
5, 6-Dimethylbenzimidazole, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 209-488-1
missing translation for 'rtecsNumber' : DD8280000
missing translation for 'tsca' : TSCA
RUO – Research Use Only