Learn More
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate, 97+%, Thermo Scientific Chemicals
Description
Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent for determination of serum iron. Reagent for the simultaneous spectrophotometric determination of iron and copper. PDT is the non-sulfonated precursor to Ferrozine (FZ), a commonly used reagent for Fe(II).
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Quantity | 1 g |
Beilstein | 5222087 |
Solubility Information | Soluble in water. |
Formula Weight | 514.46 (Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | 5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine-4,4'-disulfonic acid disodium salt hydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467293 | Certificate of Analysis | 03/12/2024 | L10607.03,L10607.06 |
A0462686 | Certificate of Analysis | 30/08/2024 | L10607.03,L10607.06 |
A0453444 | Certificate of Analysis | 20/03/2024 | L10607.03,L10607.06 |
A0420206 | Certificate of Analysis | 07/08/2023 | L10607.03,L10607.06 |
10243745 | Certificate of Analysis | 13/10/2022 | L10607.03,L10607.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.